Vikan® Pipe Brush Heads, Medium Bristles, Remco
Supplier: Remco Products
Certificates
About this item
This tube/pipe brush head has bristles along the sides as well as in the front. It is used in areas where a wider tube cleaning action is required or where the equipment has an end point that cannot be reached with a hand brush.
- Polyester bristles, polypropylene block
- Euro (hygienic) thread type
- Available in 3.0", 3.5" and 4.0" Ø
Works well as a drain brush, particularly in black. Requires a European thread handle.
Frequently Bought Together