Vikan® Tube Brushes, Medium Bristles, Remco
Fournisseur: Remco Products
Certificats
À propos de cet article
This tube brush is great for cleaning small pipes and very narrow spaces between machine parts.
- Polyester bristles, polypropylene grip
- Length: 19.9"
- Ø: 0.9"
The medium strength bristles are attached to a helically wound stainless steel shaft, which makes it strong and chemically resistant.
Frequently Bought Together