Vikan® Hook Brushes, Medium Bristles, Remco
Fournisseur: Remco Products
Certificats
À propos de cet article
Cleaning between and around pipes is made easier with the curved design of the hook brush.
- Hand grip utilizes a threaded recess on the end, allowing additional handle connections
- Polyester bristles, polypropylene grip
- Euro (hygienic) thread type
When connected to a Vikan handle, the extended hook brush allows comfortable cleaning of pipes in high or difficult to reach areas. The brush bristles are secured with noncorrosive stainless steel wires, helping increase longevity. All materials are FDA compliant, and the grip matches the bristle color for easy integration into a color-coding program.
Overall dimensions: 15.5L×2.25W×7.6"H
Frequently Bought Together