Vikan® Drain Cleaning Brushes, Stiff Bristles, Remco
Supplier: Remco Products
Certificates
About this item
The stiff bristle drain cleaning brush is used to clean small spaces between machine parts. It can also be used to clean drainage holes in sinks.
- Polyester bristles, polypropylene grip
- Length: 12.2"
- Ø: 0.6"
Short handle allows operator to exert pressure.
Frequently Bought Together