ContactUsLinkComponent
Vikan® High-Temp Detail Brush, Remco
Catalog # 76528-986
Supplier:  Remco Products
undefined
Vikan® High-Temp Detail Brush, Remco
Catalog # 76528-986
Supplier:  Remco Products
Supplier Number:  44023
Specifications
  • Description:
    High-temp detail brush, blue
  • Cat. no.:
    76528-986
Specifications
About this item
This brush can remove debris from heat sealers and other packing equipment.
  • Extra stiff bristles
  • Polypropylene block, polyester bristles
High temperature resistance also allows cleaning small gaps in grills or other equipment while they are still warm.
Overall dimensions: 8.07L×0.79W×2.36"H