Vikan® Ultra-Slim Wand Brushes, Remco
Supplier: Remco Products
Certificates
About this item
Clean where other brushes cannot reach with this ultra-slim (15 mm wide) brush designed with a scraping edge for tackling stubborn debris.
- Color-coded to support HACCP principles
- Regenerated polypropylene block, polyester bristles
- Food contact compliant
This brush features an easy-grip slim handle and a specially designed head with extra-stiff first bristles that help clean all the way to the end of narrow spaces. The brush block is made using food-contact compliant polypropylene offcuts from other Vikan products.
Overall dimensions: 23.62L×0.59W×2.36"H
Frequently Bought Together