Vikan® UST Long Handle Scrubbing Brushes, Stiff Bristles, Remco
This UST long handled hand brush with stiff bristles is effective when cleaning machines, buckets, containers, and tubs in high risk areas.
- Color-coded to support HACCP principles
- Polypropylene block, polyester bristles
The angled and circular bristle arrangement enables efficient scrubbing and washing process.
Overall dimensions: 15.5L×2.875W×2.75"H
Frequently Bought Together