Vikan® Nylon Scrapers with Threaded Handle, Remco
Fournisseur: Remco Products
Certificats
À propos de cet article
Suitable for loosening stubborn dirt like pastry, chocolate, or burnt food on smooth surfaces.
- Nylonl blade, polypropylene grip
- Euro (hygienic) thread type
The nylon blade is a good alternative to a stainless steel, and can be used on sensitive surfaces to avoid scratches on equipment and sensitive conveyor belts. The hard blade can withstand hot surfaces when used in two minute intervals. Can be used with any Vikan handle.
Frequently Bought Together