Vikan® Dustpan Broom Head with Angled Thread, Remco
Supplier: Remco Products
Certificates
About this item
Lightweight broom head can be used for sweeping both dry and wet derbis.
- Angled to faciliate sweeping close to walls, corners, and workbenches
- Polypropylene block, polyester bristles
- Euro (hygienic) thread type
An effective scraping edge helps to loosen stubborn debris. The brush pattern is designed using two bristle diameters. The outer row of bristles is softer than the middle row, which gives a very effective sweeping quality.
Broom head dimensions: 9.84L×1.18W×4.72"H
Frequently Bought Together