Vikan® Split Bristle Angled Broom Head, Remco
Fournisseur: Remco Products
Certificats
À propos de cet article
Split bristle angle head is designed to handle fine particles.
- Soft/split bristles gently sweep away powders without leaving residues
- Polypropylene block, polyester bristles
- Euro (hygienic) thread type
Made of FDA compliant materials.
Frequently Bought Together