Vikan® Hygienic Wall Bracket, Single Hook Module, Remco
Vikan® Hygienic Wall Bracket, Single Hook Module, Remco
Supplier:   Remco Products
Vikan® Hygienic Wall Bracket, Single Hook Module, Remco
Supplier:  Remco Products
Certificates
Vikan® Hygienic Wall Bracket, Single Hook Module, Remco
Vikan® Hygienic Wall Bracket, Single Hook Module, Remco
12 Options Available Below
Product Details & Documents
Single hook module is designed for storing cleaning tools that have a hanging hole.
  • Can hold products weighing up to 6.61 lbs.
  • Polypropylene/polyamide construction
The hook system is easy to disassemble for cleaning or replacement.
Product Family Options
Product Information
  • Color
    Dimensions
    Availability
    Price
  • Black
    1.63×3.07×1.89"
    Log in for availability
    Log in for availability
    Each
    Each - $21.66
    $21.66
    Each
       $21.66
    Supplier Number: 10129
  • Blue
    1.63×3.07×1.89"
    Log in for availability
    Log in for availability
    Each
    Each - $21.66
    $21.66
    Each
       $21.66
    Supplier Number: 10123
  • Brown
    1.63×3.07×1.89"
    Log in for availability
    Log in for availability
    Each
    Each - $21.66
    $21.66
    Each
       $21.66
    Supplier Number: 101266
  • Grey
    1.63×3.07×1.89"
    Log in for availability
    Log in for availability
    Each
    Each - $21.66
    $21.66
    Each
       $21.66
    Supplier Number: 101288
  • Green
    1.63×3.07×1.89"
    Log in for availability
    Log in for availability
    Each
    Each - $21.66
    $21.66
    Each
       $21.66
    Supplier Number: 10122
  • Lime green
    1.63×3.07×1.89"
    Log in for availability
    Log in for availability
    Each
    Each - $21.66
    $21.66
    Each
       $21.66
    Supplier Number: 101277
  • Orange
    1.63×3.07×1.89"
    Log in for availability
    Log in for availability
    Each
    Each - $21.66
    $21.66
    Each
       $21.66
    Supplier Number: 10127
  • Pink
    1.63×3.07×1.89"
    Log in for availability
    Log in for availability
    Each
    Each - $21.66
    $21.66
    Each
       $21.66
    Supplier Number: 10121
  • Purple
    1.63×3.07×1.89"
    Log in for availability
    Log in for availability
    Each
    Each - $21.66
    $21.66
    Each
       $21.66
    Supplier Number: 10128
  • Red
    1.63×3.07×1.89"
    Log in for availability
    Log in for availability
    Each
    Each - $21.66
    $21.66
    Each
       $21.66
    Supplier Number: 10124
  • White
    1.63×3.07×1.89"
    Log in for availability
    Log in for availability
    Each
    Each - $21.66
    $21.66
    Each
       $21.66
    Supplier Number: 10125
  • Yellow
    1.63×3.07×1.89"
    Log in for availability
    Log in for availability
    Each
    Each - $21.66
    $21.66
    Each
       $21.66
    Supplier Number: 10126