SP Bel-Art Reusable Silica Gel Desiccant Bags, Bel-Art Products, a part of SP
Fournisseur: Bel-Art Products, a Part of SP
Certificats
À propos de cet article
Convenient, easy to handle packs filled with moisture absorbing silica gel beads are available in multiple sizes to fit most desiccators.
- Durable microfiber bag securely contains the beads eliminating the risk of leakage and is unaffected by moisture
- Indicator dot changes from blue when dry to pink when moist to easily identify when recharging is needed
- Saturated gel can be regenerated by microwaving the bag for 6 minutes
- Can be regenerated repeatedly without degradation
Packaging: Supplied individually sealed in a moisture-proof pouch.
Frequently Bought Together