Optigami Build-Your-Own Cardboard Microscope Kit, Carson
Catalog # 470336-776
Supplier: CARSON OPTICAL
Specifications
- Description:Kit, build your own microscope
- W×D×H:6.6×5.6×10"
- Cat. no.:470336-776
Specifications
About this item
The Optigami will provide hours of entertainment and is a great way to nurture a child’s love of science.
- Have fun viewing the included prepared slides
- Discover things the naked eye can’t see
- Powerful 50 - 400× magnification
Experience the fun of assembling your very own microscope with the Optigami Build-Your-Own Cardboard Microscope Kit. The microscope can be used as a stand-alone, or it can become a digital microscope by attaching the smartphone platform to help capture photos or videos of your exciting finds.
Fits all phones. For specific information on your model, visit: www.carson.com/multicam.
Ordering information: Uses two AA batteries (not included).
Delivery: Includes 13 prepared slides, 2 blank microscope slides with coverslips, and (1) eye dropper.