The Wizard™ CompactDry™ Reader 2 Colony Counter
Colony counter, The Wizard™ CompactDry™ Reader 2, 3 USB ports
Catalog # CA76523-362
Supplier: HARDY DIAGNOSTICS CA
Fournisseur #: CDR2
The Wizard™ CompactDry™ Reader 2 Colony Counter
Catalog # CA76523-362
Supplier: HARDY DIAGNOSTICS CA
Jump to:
Product Details & Documents
The Wizard™ CompactDry™ Reader 2 is a fast, easy, and accurate way to read, count, and record your CompactDry™ results. Just flip the switch, select your configuration, insert your CompactDry™ plate(s), select read, and save your results.
- Accurately counts and differentiates up to 150 colonies per plate based on the CompactDry™ type in less than 4 seconds
- Stand alone system, no PC required
- Each unit includes the Hardy Data Receiver Program for file organization and storage
- Accepts individual plates or connected sets of two, three, or four plates so you can keep your dilution series together
- Unit is suited for ambidextrous operation; plates can be scanned from the left or right side
- Easy-to-operate and easy-to-clean, ergonomic with titled color touch screen for seated or standing operations
- Customizable - The unit is factory calibrated but if your samples are unique, the system has an easy-to-use calibration wizard
Digitalized plate images can be saved to USB or exported to the user's LIMS program or included software management program and there are multiple ways to report and enter reading data.
The Wizard™ has selectable modes in the configuration menu designed to read the different types of CompactDry™ plates.
Spécifications
- Désignation:Colony counter, The Wizard™ CompactDry™ Reader 2, 3 USB ports
Accessories
Informations sur le produit
- Catalog #AvailabilityPrice
- Thermal printer option for The Wizard™ CompactDry™ Reader, connects via USBThermal printer option for The Wizard™ CompactDry™ Reader, connects via USBCA76177-152Log in pour la disponibilitéEach - $2,170.00
Informations sur le produit
- Catalog #AvailabilityPrice
- Keyboard for Wizard™ CompactDry™ Reader, connects to Wizard™ by wireless USB dongleKeyboard for Wizard™ CompactDry™ Reader, connects to Wizard™ by wireless USB dongleCA76177-156Log in pour la disponibilitéEach - $295.00
Informations sur le produit
- Catalog #DimensionsAvailabilityPrice
- Replacement paper for Wizard™ CompactDry™ thermal printerReplacement paper for Wizard™ CompactDry™ thermal printerCA76177-1583¹/₈"W × 230'L, 0.5" coreLog in pour la disponibilitéEach - $10.50