Cryogenic Vials, MTC Bio
Cryogenic Vials, MTC Bio
Cryogenic Vials, MTC Bio
Cryogenic Vials, MTC Bio
Cryogenic Vials, MTC Bio
These cryogenic vials come sterile in convenient resealable bags of 50 vials.
  • Self-standing star foot design
  • Gamma sterilized
  • Rnase, Dnase, pyrogen, and endotoxin free
  • Free locking rack included with each case
Made of durable LN₂ grade polypropylene, these vials can withstand temperatures from –196 to +121 °C. The O-ring on the inner cap is made with silicone to prevent leakage.
Documents
Product Family Options
Vials, External Thread
  • Désignation
    Capacité
    Color
    Avec bouchon
    Cap Type
    Cap Material
    Sterility
    Packaging
  • Cryogenic vials
    1.00 ml
    Clear
    Oui
    Screw cap
    PP
    Sterile
    10 bags of 50
    Case of 500
    Case of 500 - 274,85 $
    274,85 $
    Case of 500
       274,85 $
     : V3801
  • Cryogenic vials
    2.00 ml
    Clear
    Oui
    Screw cap
    PP
    Sterile
    10 bags of 50
    Case of 500
    Case of 500 - 288,30 $
    288,30 $
    Case of 500
       288,30 $
     : V3802
  • Cryogenic vials
    3.00 ml
    Clear
    Oui
    Screw cap
    PP
    Sterile
    10 bags of 50
    Case of 500
    Case of 500 - 340,15 $
    340,15 $
    Case of 500
       340,15 $
     : V3803
  • Cryogenic vials
    5.00 ml
    Clear
    Oui
    Screw cap
    PP
    Sterile
    10 bags of 50
    Case of 500
    Case of 500 - 386,15 $
    386,15 $
    Case of 500
       386,15 $
     : V3805
Vials, Internal Thread
  • Désignation
    Capacité
    Color
    Avec bouchon
    Cap Type
    Cap Material
    Sterility
    Packaging
  • Cryogenic vials
    1.00 ml
    Clear
    Oui
    Screw cap
    PP
    Sterile
    10 bags of 50
    Case of 500
    Case of 500 - 255,65 $
    255,65 $
    Case of 500
       255,65 $
     : V4801
  • Cryogenic vials
    2.00 ml
    Clear
    Oui
    Screw cap
    PP
    Sterile
    10 bags of 50
    Case of 500
    Case of 500 - 288,30 $
    288,30 $
    Case of 500
       288,30 $
     : V4802
  • Cryogenic vials
    3.00 ml
    Clear
    Oui
    Screw cap
    PP
    Sterile
    10 bags of 50
    Case of 500
    Case of 500 - 341,90 $
    341,90 $
    Case of 500
       341,90 $
     : V4803
  • Cryogenic vials
    5.00 ml
    Clear
    Oui
    Screw cap
    PP
    Sterile
    10 bags of 50
    Case of 500
    Case of 500 - 386,15 $
    386,15 $
    Case of 500
       386,15 $
     : V4805
Cap Inserts
  •  
    Désignation
    Color
  • Cap insert for cryogenic vials
    Red
    Pack of 500
    Pack of 500 - 32,40 $
    32,40 $
    Pack of 500
       32,40 $
  •  
    Désignation
    Color
  • Cap insert for cryogenic vials
    Green
    Pack of 500
    Pack of 500 - 32,40 $
    32,40 $
    Pack of 500
       32,40 $
  •  
    Désignation
    Color
  • Cap insert for cryogenic vials
    Blue
    Pack of 500
    Pack of 500 - 34,85 $
    34,85 $
    Pack of 500
       34,85 $
  •  
    Désignation
    Color
  • Cap insert for cryogenic vials
    Assorted
    Pack of 500
    Pack of 500 - 32,40 $
    32,40 $
    Pack of 500
       32,40 $
  •  
    Désignation
    Color
  • Cap insert for cryogenic vials
    White
    Pack of 500
    Pack of 500 - 32,40 $
    32,40 $
    Pack of 500
       32,40 $
  •  
    Désignation
    Color
  • Cap insert for cryogenic vials
    Yellow
    Pack of 500
    Pack of 500 - 32,40 $
    32,40 $
    Pack of 500
       32,40 $
Storage
  •  
    Désignation
  • 50-place locking rack for cryogenic vials
    Pack of 4
    Pack of 4 - 98,70 $
    98,70 $
    Pack of 4
       98,70 $