Microsart® @filter Membrane Filter Funnels
Supplier:   Sartorius
Microsart® @filter Membrane Filter Funnels
Supplier:  Sartorius
Certificates
Microsart® @filter Membrane Filter Funnels
27 Options Available Below
Product Details & Documents
These filter units with PP funnel and base and cellulose nitrate membrane are developed for the detection and enumeration of microorganisms in pharmaceuticals, cosmetics, food, beverages, water and other liquids. All components of the filtration system must comply with international guidelines, such as USP, EP or ISO standards.
  • No liquid remains after filtration, no need for rinsing
  • Click-Fit closure, no risk of leakage
  • Adaptable on Combisart®
  • Transparent funnel material with marked graduations
Microsart® Base 47 mm is the perfect addition to existing Combisart® and Microsart® Combi.jet stainless steel manifolds. The Microsart® funnel dispenser guarantees secure removal of single, sterile Microsart® @filter units.
Capacity 100 ml: graduations at 20, 50 and 100 ml.
Capacity 250 ml: graduations at 50, 100, 200 and 250 ml.
Specifications for Product Family
Spécifications
  • Catalog No:
  • CA76517-854
  • CA76517-856
  • CA76517-860
  • CA76517-862
  • CA76517-866
  • CA76517-882
  • CA76517-886
  • CA76517-906
  • CA76517-870
  • CA76517-874
  • CA76517-876
  • CA76517-880
  • CA76517-894
  • CA76517-890
  • CA76517-896
  • CA76517-852
  • CA76517-858
  • CA76517-864
  • CA76517-884
  • CA76517-904
  • CA76517-908
  • CA76517-910
  • CA76517-892
  • CA76517-888
  • CA76517-878
  • CA76517-872
  • CA76517-868
  • Pre-filter:
  • 40 mm
  • 40 mm
  • 40 mm
  • 40 mm
  • 40 mm
  • 40 mm
  • 40 mm
  • 40 mm
  • 40 mm
  • 40 mm
  • 40 mm
  • 40 mm
  • 40 mm
  • 40 mm
  • 40 mm
  • 40 mm
  • 40 mm
  • 40 mm
  • 40 mm
  • 40 mm
  • 40 mm
  • 40 mm
  • 40 mm
  • 40 mm
  • 40 mm
  • 40 mm
  • 40 mm
  • Filtration area:
  • 13.2 cm²
  • 13.2 cm²
  • 13.2 cm²
  • 13.2 cm²
  • 13.2 cm²
  • 13.2 cm²
  • 13.2 cm²
  • 13.2 cm²
  • 13.2 cm²
  • 13.2 cm²
  • 13.2 cm²
  • 13.2 cm²
  • 13.2 cm²
  • 13.2 cm²
  • 13.2 cm²
  • 13.2 cm²
  • 13.2 cm²
  • 13.2 cm²
  • 13.2 cm²
  • 13.2 cm²
  • 13.2 cm²
  • 13.2 cm²
  • 13.2 cm²
  • 13.2 cm²
  • 13.2 cm²
  • 13.2 cm²
  • 13.2 cm²
Spécifications
Product Family Options
Funnel with lid
Informations sur le produit
  • Désignation
    Capacité
    Taille de pore
    Ø
    Membrane
    Packaging
    Availability
    Price
  • Membrane colour | Grid colour: white/black
    100 ml
    0.45 µm
    47 mm
    Cellulose nitrate
    3 trays of 8
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 24
    Pack of 24 - $206.94
    $206.94
    Pack of 24
       $206.94
    Supplier Number: 16D011006TG
    Spécifications:
    More
    Specifications
    Cat. No.
    CA76517-854
    Pre-filter
    40 mm
    Filtration area
    13.2 cm²
  • Membrane colour | Grid colour: white/black
    100 ml
    0.2 µm
    47 mm
    Cellulose nitrate
    27 per pack
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 27
    Pack of 27 - $311.09
    $311.09
    Pack of 27
       $311.09
    Supplier Number: 16D011007ACG
    Spécifications:
    More
    Specifications
    Cat. No.
    CA76517-856
    Pre-filter
    40 mm
    Filtration area
    13.2 cm²
  • Membrane colour | Grid colour: white/black
    100 ml
    0.2 µm
    47 mm
    Cellulose nitrate
    3 trays of 8
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 24
    Pack of 24 - $205.92
    $205.92
    Pack of 24
       $205.92
    Supplier Number: 16D011007TG
    Spécifications:
    More
    Specifications
    Cat. No.
    CA76517-860
    Pre-filter
    40 mm
    Filtration area
    13.2 cm²
  • Membrane colour | Grid colour: white/black
    100 ml
    0.45 µm
    47 mm
    Cellulose nitrate
    27 individually packed
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $267.03
    $267.03
    Each
       $267.03
    Supplier Number: 16D0110H6ACG
    Spécifications:
    More
    Specifications
    Cat. No.
    CA76517-862
    Pre-filter
    40 mm
    Filtration area
    13.2 cm²
  • Membrane colour | Grid colour: white/black
    100 ml
    0.45 µm
    47 mm
    Cellulose nitrate
    3 trays of 8
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 24
    Pack of 24 - $206.94
    $206.94
    Pack of 24
       $206.94
    Supplier Number: 16D0110H6TG
    Spécifications:
    More
    Specifications
    Cat. No.
    CA76517-866
    Pre-filter
    40 mm
    Filtration area
    13.2 cm²
  • Membrane colour | Grid colour: black/white
    100 ml
    0.45 µm, high flow
    47 mm
    Cellulose nitrate
    27 per pack
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 27
    Pack of 27 - $267.03
    $267.03
    Pack of 27
       $267.03
    Supplier Number: 16D0310H6ACG
    Spécifications:
    More
    Specifications
    Cat. No.
    CA76517-882
    Pre-filter
    40 mm
    Filtration area
    13.2 cm²
  • Membrane colour | Grid colour: gray/white
    100 ml
    0.45 µm
    47 mm
    Cellulose nitrate
    3 trays of 8
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 24
    Pack of 24 - $206.94
    $206.94
    Pack of 24
       $206.94
    Supplier Number: 16D0310H6TG
    Spécifications:
    More
    Specifications
    Cat. No.
    CA76517-886
    Pre-filter
    40 mm
    Filtration area
    13.2 cm²
  • Membrane colour | Grid colour: white/without grid
    100 ml
    0.45 µm
    47 mm
    Regenerated cellulose
    24 per pack
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 24
    Pack of 24 - $206.94
    $206.94
    Pack of 24
       $206.94
    Supplier Number: 16D051006TG
    Spécifications:
    More
    Specifications
    Cat. No.
    CA76517-906
    Pre-filter
    40 mm
    Filtration area
    13.2 cm²
  • Membrane colour | Grid colour: white/black
    250 ml
    0.2 µm
    47 mm
    Cellulose nitrate
    18 per pack
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 18
    Pack of 18 - $195.82
    $195.82
    Pack of 18
       $195.82
    Supplier Number: 16D012507ACF
    Spécifications:
    More
    Specifications
    Cat. No.
    CA76517-870
    Pre-filter
    40 mm
    Filtration area
    13.2 cm²
  • Membrane colour | Grid colour: white/black
    250 ml
    0.2 µm
    47 mm
    Cellulose nitrate
    2 trays of 8
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 16
    Pack of 16 - $145.89
    $145.89
    Pack of 16
       $145.89
    Supplier Number: 16D012507TF
    Spécifications:
    More
    Specifications
    Cat. No.
    CA76517-874
    Pre-filter
    40 mm
    Filtration area
    13.2 cm²
  • Membrane colour | Grid colour: white/black
    250 ml
    0.45 µm, high flow
    47 mm
    Cellulose nitrate
    18 per pack
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 18
    Pack of 18 - $195.82
    $195.82
    Pack of 18
       $195.82
    Supplier Number: 16D0125H6ACF
    Spécifications:
    More
    Specifications
    Cat. No.
    CA76517-876
    Pre-filter
    40 mm
    Filtration area
    13.2 cm²
  • Membrane colour | Grid colour: white/black
    250 ml
    0.45 µm
    47 mm
    Cellulose nitrate
    2 trays of 8
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 16
    Pack of 16 - $153.34
    $153.34
    Pack of 16
       $153.34
    Supplier Number: 16D0125H6TF
    Spécifications:
    More
    Specifications
    Cat. No.
    CA76517-880
    Pre-filter
    40 mm
    Filtration area
    13.2 cm²
  • Membrane colour | Grid colour: gray/white
    250 ml
    0.45 µm, high flow
    47 mm
    Cellulose nitrate
    10 per pack
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 16
    Pack of 16 - $1,585.02
    $1,585.02
    Pack of 16
       $1,585.02
    Supplier Number: 16D0325H6BS
    Spécifications:
    More
    Specifications
    Cat. No.
    CA76517-894
    Pre-filter
    40 mm
    Filtration area
    13.2 cm²
  • Membrane colour | Grid colour: black/white
    250 ml
    0.45 µm, high flow
    47 mm
    Cellulose nitrate
    18 per pack
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 18
    Pack of 18 - $195.82
    $195.82
    Pack of 18
       $195.82
    Supplier Number: 16D0325H6ACF
    Spécifications:
    More
    Specifications
    Cat. No.
    CA76517-890
    Pre-filter
    40 mm
    Filtration area
    13.2 cm²
  • Membrane colour | Grid colour: gray/white
    250 ml
    0.45 µm
    47 mm
    Cellulose nitrate
    2 trays of 8
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 16
    Pack of 16 - $147.46
    $147.46
    Pack of 16
       $147.46
    Supplier Number: 16D0325H6TF
    Spécifications:
    More
    Specifications
    Cat. No.
    CA76517-896
    Pre-filter
    40 mm
    Filtration area
    13.2 cm²
Funnel without lid
Informations sur le produit
  • Désignation
    Capacité
    Taille de pore
    Ø
    Membrane
    Packaging
    Availability
    Price
  • Membrane colour | Grid colour: white/black
    100 ml
    0.45 µm
    47 mm
    Cellulose nitrate
    6 bags of 10
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 60
    Pack of 60 - $466.35
    $466.35
    Pack of 60
       $466.35
    Supplier Number: 16D011006BL
    Spécifications:
    More
    Specifications
    Cat. No.
    CA76517-852
    Pre-filter
    40 mm
    Filtration area
    13.2 cm²
  • Membrane colour | Grid colour: white/black
    100 ml
    0.2 µm
    47 mm
    Cellulose nitrate
    6 bags of 10
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 60
    Pack of 60 - $442.58
    $442.58
    Pack of 60
       $442.58
    Supplier Number: 16D011007BL
    Spécifications:
    More
    Specifications
    Cat. No.
    CA76517-858
    Pre-filter
    40 mm
    Filtration area
    13.2 cm²
  • Membrane colour | Grid colour: white/black
    100 ml
    0.45 µm
    47 mm
    Cellulose nitrate
    6 bags of 10
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 60
    Pack of 60 - $442.58
    $442.58
    Pack of 60
       $442.58
    Supplier Number: 16D0110H6BL
    Spécifications:
    More
    Specifications
    Cat. No.
    CA76517-864
    Pre-filter
    40 mm
    Filtration area
    13.2 cm²
  • Membrane colour | Grid colour: gray/white
    100 ml
    0.45 µm
    47 mm
    Cellulose nitrate
    6 bags of 10
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 60
    Pack of 60 - $515.62
    $515.62
    Pack of 60
       $515.62
    Supplier Number: 16D0310H6BL
    Spécifications:
    More
    Specifications
    Cat. No.
    CA76517-884
    Pre-filter
    40 mm
    Filtration area
    13.2 cm²
  • Membrane colour | Grid colour: white/without grid
    100 ml
    0.45 µm
    47 mm
    Regenerated cellulose
    60 per pack
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 60
    Pack of 60 - $515.62
    $515.62
    Pack of 60
       $515.62
    Supplier Number: 16D051006BL
    Spécifications:
    More
    Specifications
    Cat. No.
    CA76517-904
    Pre-filter
    40 mm
    Filtration area
    13.2 cm²
  • Membrane colour | Grid colour: white/no grid
    100 ml
    0.45 µm
    47 mm
    Polycarbonate
    60 per pack
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 60
    Pack of 60 - $457.63
    $457.63
    Pack of 60
       $457.63
    Supplier Number: 16D061006BL
    Spécifications:
    More
    Specifications
    Cat. No.
    CA76517-908
    Pre-filter
    40 mm
    Filtration area
    13.2 cm²
  • Membrane colour | Grid colour: white/no grid
    250 ml
    0.2 µm
    47 mm
    Polycarbonate
    48 per pack
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 48
    Pack of 48 - $532.66
    $532.66
    Pack of 48
       $532.66
    Supplier Number: 16D062507BK
    Spécifications:
    More
    Specifications
    Cat. No.
    CA76517-910
    Pre-filter
    40 mm
    Filtration area
    13.2 cm²
  • Membrane colour | Grid colour: gray/white
    250 ml
    0.45 µm
    47 mm
    Cellulose nitrate
    6 bags of 8
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 48
    Pack of 48 - $434.51
    $434.51
    Pack of 48
       $434.51
    Supplier Number: 16D0325H6BK
    Spécifications:
    More
    Specifications
    Cat. No.
    CA76517-892
    Pre-filter
    40 mm
    Filtration area
    13.2 cm²
  • Membrane colour | Grid colour: gray/white
    250 ml
    0.65 µm
    47 mm
    Cellulose nitrate
    6 bags of 8
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 48
    Pack of 48 - $434.51
    $434.51
    Pack of 48
       $434.51
    Supplier Number: 16D032505BK
    Spécifications:
    More
    Specifications
    Cat. No.
    CA76517-888
    Pre-filter
    40 mm
    Filtration area
    13.2 cm²
  • Membrane colour | Grid colour: white/black
    250 ml
    0.45 µm
    47 mm
    Cellulose nitrate
    6 bags of 8
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 48
    Pack of 48 - $434.51
    $434.51
    Pack of 48
       $434.51
    Supplier Number: 16D0125H6BK
    Spécifications:
    More
    Specifications
    Cat. No.
    CA76517-878
    Pre-filter
    40 mm
    Filtration area
    13.2 cm²
  • Membrane colour | Grid colour: white/black
    250 ml
    0.2 µm
    47 mm
    Cellulose nitrate
    6 bags of 8
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 48
    Pack of 48 - $434.51
    $434.51
    Pack of 48
       $434.51
    Supplier Number: 16D012507BK
    Spécifications:
    More
    Specifications
    Cat. No.
    CA76517-872
    Pre-filter
    40 mm
    Filtration area
    13.2 cm²
  • Membrane colour | Grid colour: white/black
    250 ml
    0.45 µm
    47 mm
    Cellulose nitrate
    6 bags of 8
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 48
    Pack of 48 - $434.38
    $434.38
    Pack of 48
       $434.38
    Supplier Number: 16D012506BK
    Spécifications:
    More
    Specifications
    Cat. No.
    CA76517-868
    Pre-filter
    40 mm
    Filtration area
    13.2 cm²