Microsart® e.jet Pump
Microsart® e.jet fluid pump, 4 Nl/min., 100/240 V (US plug)
  CA76517-796
 :   Sartorius
 :  166MP4
Microsart® e.jet Pump
  CA76517-796
 :  Sartorius
Microsart® e.jet Pump
Microsart® e.jet Pump
Microsart® e.jet fluid pump, 4 Nl/min., 100/240 V (US plug)
$2,738.81
Each
The Microsart® e.jet is a laboratory vacuum pump which creates vacuum and concomitantly transfers the filtered liquid to waste.
  • For vacuum filtration in microbiological analysis
Microsart® e.jet is ideal for sample preparation in microbiological analysis achieving a trans membrane pressure of 600 mbar and a flow rate of 4.0 Nl/min (4.0 Normlitre water displacement by air in 1 minute). Constant flow rates and a defined maximum vacuum guarantees smooth and reliable filtration.
  • Désignation:
    Microsart® e.jet fluid pump, 4 Nl/min.
  • Electrical:
    100/240 V (US plug)
Accessories
  •  
    Description
    For Use With
  • CA76517-940
    Quick connection set, 2 nipples (POM) on R³/₈" male thread and 2 couplings (PSU) on DN 10 hose nipple
    Microsart® e.jet pump
    Pack of 2 - $51.60