Rubbermaid® Economy Wet Mopping Pad, Microfiber, Essendant
Catalog # 76500-188
Supplier: Janitorial Supplies
Specifications
- Color:Blue
- Material:Microfiber
- Size:18×5"
- Cat. no.:76500-188
Specifications
About this item
This damp room mop is made from extremely thin synthetic fibers that are non-abrasive and lint-free, and remove more dust, dirt, and microbes than conventional cleaning products.
- Made from extremely thin nylon/polyester blend fibers that are non-abrasive and lint-free
- Bleach-safe
The density of the material makes the cleaning pad more absorbent than conventional cotton looped-end mops. These Rubbermaid mop heads will withstand hundreds of launderings for long product usage life.
The Rubbermaid Commercial Products Microfiber Floor Mop is designed for use with the Rubbermaid FGQ55900 standard Quick-Connect frame (sold separately).