Rubbermaid® Invader Side-Gate Wood Wet-Mop Handle, Essendant
Catalog # 76500-124
Supplier: Janitorial Supplies
Specifications
- Color:Natural
- Material:Wood
- Cat. no.:76500-124
Specifications
About this item
Easy-change, side-gate design allows users to quickly slide mop heads on or off.
- Easily remove and replace mop heads
- Side gate makes wet mop head connection easy
- Handle length: 60"
- Handle diameter: 1"
Adjustable thumb wheel enables tight and secure clamp down. Solvent- and acid-resistant frame allows use with all commercial floor stripping agents. For use with 1" headband mop heads.