Rubbermaid® Pulse Microfiber Spray Mop System, Essendant
Catalog # 76500-110
Supplier: Janitorial Supplies
Specifications
- Description:Pulse Microfiber spray mop system
- Material:Plastic handle, Microfiber mop head
- Cat. no.:76500-110
Specifications
About this item
Clean more square feet in less time with this reliable mop system.
- Red ergonomic trigger handle dispenses three streams of cleaning solution with each press
- Lightweight construction; only 4 lbs. when fully loaded with solution
- High-capacity refillable reservoir holds 21 oz. of solution to clean up to 850 sq. ft. of floor space
- Reservoir windows allow easy monitoring of fluid level
- 17" wide mop head
- 52" handle length, 56" overall length
Industry-best microfiber, on-board reservoir, and user-controlled release of solution results in faster, easier, and more effective cleaning.
System includes one trigger handle with fluid reservoir, a 17" Quick-Connect frame, and two 18" Hygen™ Damp mop pads. Also compatible with Rubbermaid® 11" and 24" Quick-Connect frames and pads.