Boardwalk® Lie-Flat Screw-In Mop Handle, Essendant
76500-102
- Color:Natural
- Description:Lie-Flat Screw-in mop handle
- Material:Wood, lacquered
- Cat. no.:76500-102
Use with bolt head style lie-flat mop head.
- Lacquered wood construction
- Natural finish
- 1.125" handle diameter
- 60" handle length
Metal-threaded connector is countersunk and riveted.