ContactUsLinkComponent
DirectPlate™ JM109 Chemically Competent Escherichia coli Cells, Intact Genomics
DirectPlate™ JM109 Chemically Competent Escherichia coli Cells, Intact Genomics
Catalog # 104013-340
Supplier:  Intact Genomics
DirectPlate™ JM109 Chemically Competent Escherichia coli Cells, Intact Genomics
Catalog # 104013-340
Supplier:  Intact Genomics
Supplier Number:  1016-36
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Size:
    36×50 µl
  • Cat. no.:
    104013-340
Specifications
About this item
DirectPlate™ JM109 chemically competent Escherichia coli cells are a perfect choice for researchers looking to simplify their transformation workflow by eliminating heat shock, lengthy incubations, and time-consuming outgrowth procedures. Simply mix and directly plate! 
  • No heat shock
  • Save time (3 minutes transformation instead of a 90 minute transformation)
  • High transformation efficiency
DirectPlate™ JM109 chemically competent Escherichia coli cells provide higher transformation efficiency than any competitor‘s similar product and are suitable for high-efficiency transformation in a wide variety of applications such as cloning and sub-cloning. Intact Genomics (ig®) DirectPlate™ competent cells offer simple, fast, and robust results for your DNA transformation needs. Genotype: endA1, recA1, gyrA96, thi, hsdR17 (rk–, mk+), relA1, supE44, Δ( lac-proAB), [F´ traD36, proAB, laqIqZΔM15].
Delivery: All our products are shipped with dry ice via FedEx overnight delivery. US orders received by 1:00 pm (Central time, Monday-Thursday) will typically be shipped on the same day in the US. Orders after 1:00 PM will be shipped on the next business day. Orders received on Friday will be shipped on the following Monday.
The shipping and dry ice packaging cost is $45 for your entire order in the 48 contiguous United States.