Intact Genomics C58C1 chemically competent Agrobacterium cells are useful for transgenic operations that involve potatoes, astragalus, rosaceae, tobacco and other plants.
- High transformation efficiencies
- Especially useful for cDNA gDNA library construction
Intact Genomics (ig®) C58C1 chemically competent Agrobacterium cells are optimized for the highest transformation efficiency and are useful for various applications. The chromosomal background of C58C1 is C58. C58 is cured of the Ti plasmid pTiC58 resulting in C58C1. C58C1 competent cells may be useful for transgenic operations that involve Arabidopsis and other plants. This Agrobacterium strain is streptomycin and rifampicin resistant.
The shipping and dry ice packaging cost is $45 for your entire order in the 48 contiguous United States.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.