About this item
Intact Genomics JM109 chemically competent Escherichia coli cells are suitable for high efficiency transformation in a wide variety of applications such as cloning and sub-cloning.
- recA– and endA– to minimize recombination and improve the quality of plasmid DNA
- Contains F’ episome for blue/white screening
JM109 is a K strain that is recA– and endA– to minimize recombination and improve the quality of plasmid DNA. Suitable for high efficiency transformation in a wide variety of applications. Genotype: endA1, recA1, gyrA96, thi, hsdR17 (rk–, mk+), relA1, supE44, Δ( lac-proAB), [F´ traD36, proAB, laqIqZΔM15].
The shipping and dry ice packaging cost is $45 for your entire order in the 48 contiguous United States.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.