ContactUsLinkComponent
Endonuclease IV (NFO) 5,000 Units, Intact Genomics
  104013-272
 :  
undefined
Endonuclease IV (NFO) 5,000 Units, Intact Genomics
  104013-272
 :  3425
 :  
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
  • Size:
    10U/µl
  • Cat. no.:
    104013-272
Endonuclease IV (Nfo) from Escherichia coli is a 32 kD metalloprotein that aids in the repair of damaged DNA. The enzyme functions both as an apurinic/apyrimidinic nuclease (1) and as a 3′-terminal di-esterase (1-4).
  • No detectable exonuclease or DNA N-glycosylase activity
  • Useful for single cell gel electrophoresis, alkaline elution, and alkaline unwinding
Its 3′-terminal di-esterase activity is important in the repair of DNA strand breaks generated by oxidation and ionic radia-tion (2, 3). In such events, the strand breaks terminate with either a 3′ phosphate or a deoxyribose fragment, preventing repair by DNA polymerase I or DNA ligase. Endonuclease IV removes the blocking groups, leaving a free 3′-hydroxyl terminus. This enzyme does not have detectable associated exonuclease or DNA N-glycosylase activity (1).
 : All our products are shipped with dry ice via FedEx overnight delivery. US orders received by 1:00 pm (Central time, Monday-Thursday) will typically be shipped on the same day in the US. Orders after 1:00 PM will be shipped on the next business day. Orders received on Friday will be shipped on the following Monday.
The shipping and dry ice packaging cost is $45 for your entire order in the 48 contiguous United States.