Rubbermaid® Economy Cut-End Cotton Wet Mop Head, Essendant
76500-372
- Color:White
- Description:Economy cut-end wet mop head
- Material:Cotton
- Cat. no.:76500-372
Ideal for general purpose cleaning, this all-purpose mop makes cleaning a breeze with 4-ply cotton fibers made from up to 100% recycled post-industrial cotton.
- Cotton mop for general, all-purpose cleaning
- Recycled content post-industrial cotton
- Shipped as compact mop packages
- 1" headband
Non-launderable, these mops require bucket/wringer use only.