Rubbermaid® HYGEN Dry Dusting Mop Head with Fringe, Essendant
Catalog # 76500-302
Supplier: Janitorial Supplies
Specifications
- Color:Green
- Description:HYGEN dry dusting mop head with fringe
- Material:Microfiber
- Size:36"
- Cat. no.:76500-302
Specifications
About this item
100% microfiber dust mop collects and holds large dust particles, debris, and dirt mechanically and electrostatically for superior dusting results.
- 100% microfiber dust mop
- Microfibers ensure outstanding dust and microbe removal from all types of surfaces
- Advanced hook-and-loop backing
Microfibers ensure outstanding dust and microbe removal from all types of cleaning surfaces. Advanced hook-and-loop backing holds-up under harsh laundering conditions and attaches pads more securely.