LYSOL® Disinfecting Wipes, Lemon and Lime Blossom, Essendant
76500-264
- Color:White
- L×W:7×7.25"
- Cat. no.:76500-264
- Packaging:80 wipes/canister
Each thick, pre-moistened wipe kills 99.9% of viruses and bacteria on hard non-porous surfaces when used as directed. Ideal for cleaning, disinfecting, and deodorizing.
- Kills the cold and flu viruses
- Ideal to clean, disinfect, and deodorize
- Remove more than 95% of allergens, including pet dander, dust mites, and pollen
- Safe to use on electronics including smartphones, tablets, and remote controls
Wipes are effective against SARS-CoV-2, the virus that causes COVID-19, in 15 seconds when used as directed on hard, non-porous surfaces. Wipes also remove more than 95% of allergens, including pet dander, dust mites, and pollen. Use on kitchen counters, appliances, stove tops, cabinets, door knobs, light switches, floors, garbage cans, toilet exteriors, bathtubs, and sinks. Also safe to use on electronics including smartphones, tablets, and remote controls.