CrossLab Capillary and Tubing Kits for HPLC, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
Certificates
About this item
A complete set of system capillaries and fittings to efficiently ramp up usage of LC instruments.
- Carefully selected capillary dimensions (inner diameter and length) to minimize extra-column volume and optimize separation efficiency
- Peace of mind of having everything needed to make necessary connections on your system
- InfinityLab Quick Connect and Quick Turn fittings in the InfinityLab capillary kits ensure ease of use and zero dead-volume column connection
Agilent InfinityLab capillary kits are complete sets of system capillaries, including InfinityLab Quick-Connect and Quick Turn fittings, to efficienctly ramp up usage of InfinityLab LC Series instruments.
Agilent capillary/fitting starter kits contain the most widely used connection capillaries, fittings, and tools needed for older LC systems. Capillary dimensions are optimized to minimize extra-column volume to ensure highest separation efficiency.
Frequently Bought Together