VWR® Screw Caps for 4 ml Screw-Thread Vials
Frequently Bought Together
B352 Specimen Bottles and Caps, Simport Scientific
: Simport Scientific
VWR® Manual Crimping and Decapping Tools
: VWR International
Plastic Vials, Caps and Convenience Kits for LCMS and HPLC
: MicroSolv Technology Corp
VWR® Snap Caps for 11 mm Snap Ring Vials, Wide Opening
: VWR International
VWR® Spectrosil Spectrophotometer Cells
: VWR International
VWR® Packer Style Bottles, Amber, Wide Mouth
: VWR International
VWR® Culture Tubes, Disposable, with Screw-Cap Finish, Borosilicate Glass
: VWR International
MSCERT KIT 2 ML CLR SCR 9MM PK100
: Thermo Scientific
Cryoscope Standards, Advanced Instruments
: Advanced Instruments
Acetone ACS
: INNOVA CHEMICALS CA
High Recovery Vials and Inserts, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD) CA
RSA™ and RSA-Pro™ Autosampler Vials, Caps and Convenient Kits
: MicroSolv Technology Corp