Disposable Polystyrene Cells for UV-Vis and UV-Vis-NIR Systems, Agilent Technologies
Fournisseur: AGILENT TECHNOLOGIES, INC (CSD) CA
Certificats
À propos de cet article
Disposable plastic standard or micro cells are economical alternatives to quartz or glass cells.
- Eliminate the risk of cross-contamination with traditional quartz reusable cuvettes
- Improve productivity and reduce cost - eliminate the need for cleaning cuvettes for re-use
- Universal - compatible with virtually all UV/VIS systems without adapters
- Available in 2 sizes, both with 10 mm path length, 3.5 ml (standard) and 1.5 ml (semi-micro)
- Usable over wavelength range from 340 - 900 nm
- Packed in 5 styrofoam trays, each tray holding 100 cells
- Not recommended for use at elevated temperatures
Disposable plastic cuvettes are economical alternatives to quartz or glass rectangular cells. Both standard and micro cells are compatible with most UV-Vis systems. They eliminate the need for cleaning, improving productivity and reducing laboratory costs. They also reduce cross-contamination risks.
Packaging: Packed in 5 styrofoam trays, each tray holding 100 cells.
Caution: Not recommended for use at elevated temperatures.
Frequently Bought Together