Electronic Crimpers & Decappers, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
Certificates
About this item
A choice of handheld electronic crimpers, decappers, and related accessories, including the A-Line crimper and decapper.
- OLED screen with multilanguage capabilities
- Brushless motor increases lifetime and reduces battery drain
- Crimping capability reporting
- Lightweight to reduce hand strain
Agilent 11 mm and 20 mm handheld electronic vial crimpers deliver tight, reproducible seals every time. Slim, adjustable steel jaws fit around closely spaced crimp vials, enabling you to crimp vials directly in vial trays or vial racks. A-Line vial crimpers offer improved crimping performance, an OLED screen in multiple languages, and a brushless motor promoting longer usable life. A-Line electronic vial decappers remove crimp caps quickly, and are designed for labs that recycle or re-use vials.
Frequently Bought Together