Disintegration Baskets, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
Disintegration Baskets, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
Certificates
Jump to:
À propos de cet article
Agilent offers disintegration baskets for the 100 automated disintegration apparatus.
10 Options Available Below
Product Details & Documents
Agilent offers disintegration baskets for the 100 automated disintegration apparatus.
Product Family Options
Informations sur le produit
- DésignationAvailabilityPrice
- Supplier Number: 37-4010
- Basket, 1-tube, 44 mm OD, 10-mesh, 1905 µmDiscontinued-Supplier Number: 37-3040
- Supplier Number: 37-2030
- Supplier Number: 37-3030
- Basket, 3-tube, 38 mm OD, 10-mesh, 1905 µm, includes certificateLog in pour la disponibilitéEach$1,530.00Supplier Number: 37-3030V
- Supplier Number: 37-3001
- Supplier Number: 37-2001
- Basket, 6-tube, 10-mesh, 1905 µm (USP), includes certificateLog in pour la disponibilitéEach$982.00Supplier Number: 37-2001V
- Basket, 6-tube, 10-mesh, 1905 µm (USP), includes certificateLog in pour la disponibilitéEach$989.00Supplier Number: 37-3001V
- Supplier Number: 37-2010
- Supplier Number: 37-3010
- Basket, 6-tube, 40-mesh, 381 µm, with screened coverDiscontinued-Supplier Number: -