Detector Lamps for Waters LC Systems for HPLC, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
Certificates
About this item
Lamps for selected Waters HPLC systems compatible with a wide variety of detectors.
- High output stability and intensity for extended detection capabilities and improved qualification at trace level
- Guaranteed lifetime of 2000 hours
Agilent lamps for selected Waters HPLC systems are designed and manufactured to be compatible with a wide variety of detectors, including multiple wavelength detectors (MWD) and photodiode array detectors (PDA).
All the lamps adhere to the tightest specifications for consistent quality and reproducible performance over the entire lifetime of the lamps.
Frequently Bought Together