CrossLab Integrated Autosampler (I-AS) Supplies for ICP-MS, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
Certificates
About this item
Integrated autosampler (I-AS) supplies for ICP-MS for ultratrace analysis on Agilent ICP-MS systems.
- These supplies including replacement tubing kits for in-built peristaltic pump and sample probes, which may be integrated with PFA concentric nebulizers including MicroFlow and C-Flow
- Covered autosampler for use with the Agilent single and triple quadrupole ICP-MS
- No metal or drive mechanism moves over sample vials
- Continuous flowing rinse port for probe wash
- Choice of sample tray arrangements, small and large sample volumes accommodated
- Available with 3 × 100 ml standard or wash bottles
- Fully automated QA/QC control procedures available using the optional Intelligent Sequencing Software
- Designed for all semiconductor applications and any ultratrace analysis
Agilent offers the fast, flexible, and high-precision integrated autosampler (I-AS) specifically for ultratrace analysis on Agilent ICP-MS systems. The I-AS is ideal for semiconductor applications. In addition, a range of QA/QC control procedures, such as EPA protocols, can be automated using the optional Intelligent Sequencing software.
Frequently Bought Together