CrossLab Graphite Furnace AAS Graphite Shrouds, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
Certificates
About this item
Spare graphite shroud to ensure optimum performance with maximum tube life.
- Keeping a spare graphite shroud on-hand helps you maximize uptime
- Good thermal and gas isolation ensures optimum performance and maximizes tube life
- High purity graphite reduces spurious absorption from trace contaminants and improves signal-to-noise ratio
Graphite shrouds for graphite furnace Atomic Absorption. Regular replacement of the shroud is recommended. A shroud in good condition will help ensure optimum performance and maximize tube life.
Frequently Bought Together