CrossLab Autosampler Sampling Capillaries, Agilent Technologies
CrossLab Autosampler Sampling Capillaries, Agilent Technologies
Certificates
CrossLab Autosampler Sampling Capillaries, Agilent Technologies
CrossLab Autosampler Sampling Capillaries, Agilent Technologies
3 Options Available Below
Product Details & Documents
Dispensing capillary assemblies forAgilent PSD 100/120 Programmable Sample Dispenser.
  • Works well with solvent and acid-resistant sampling components
  • PTFE capillary tips and capillaries reduce contamination
  • Easy replacement by the user
Agilent provides a range of supplies for the PSD 100/120 Programmable Sample Dspenser used with Agilent GFAAS systems, including sample vials, beakers, adapters, dispensing capillary assemblies, dispensing syringe and replacement plungers.​
Dispensing capillary assemblies are made from PTFE tubing, making them ideal for collecting and dispensing a variety of corrosive matrices and organic solutions.
Product Family Options
Informations sur le produit
  • Désignation
    Availability
    Price
  • Capillary assembly, PTFE, 0.022 in Int. Ø, 0.05" Ext. Ø
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 5
    Pack of 5 - $483.00
    $483.00
    Pack of 5
       $483.00
    Supplier Number: 9910115100
  • Capillary assembly, PTFE, 0.022 in Int. Ø, 0.05" Ext. Ø
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 5
    Pack of 5 - $831.00
    $831.00
    Pack of 5
       $831.00
    Supplier Number: 9910032300
  • Kit 5 rinse bottle capillaries, PTFE, 0.03" Int. Ø, 0.062" Ext. Ø
    Log in pour la disponibilité
    Log in pour la disponibilité
    Kit of 1
    Kit of 1 - $640.00
    $640.00
    Kit of 1
       $640.00
    Supplier Number: 9910034200