CrossLab Column Nuts for GC
CrossLab Column Nuts for GC
CrossLab Column Nuts for GC
Certificates
CrossLab Column Nuts for GC
CrossLab Column Nuts for GC
25 Options Available Below
Product Details & Documents
Column nuts come in ¹/₁₆" stainless steel, ¹/₈" and ¹/₄" brass, and new self tightening nuts.
  • No retightening means less wasted time
  • Finger-tight design lets anyone make consistent quality connections without tools
  • Low-torque seal prevents ferrules from sticking or crumbling
  • Lower column bleed prolongs column life
One of the most basic tasks in GC, installing the column, can be challenging. The GC oven is a dark, small space, and column installation uses tiny parts that need to be assembled with precision. Installation of the column into both the inlet and detector must be done properly to get accurate and reproducible results. To facilitate good column installation, Agilent offers a selection of capillary column nuts for GC fittings. The latest innovation, the Self Tightening column nut, prevents leaking and the need to retighten when using graphite/polyimide blend ferrules, which are recommended for use with oxygen sensitive detectors such as ECD or mass spectrometry.
Product Family Options
Informations sur le produit
  • Désignation
    Matière
    Max. temp.
    Availability
    Price
  • Ferrule, ¹/₈"
    PTFE
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $32.50
    $32.50
    Each
       $32.50
    Supplier Number: 0100-1365
  • Front ferrule, ¹/₁₆" (equivalent to J&W 309-0001)
    PTFE
    250 °C
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 10
    Pack of 10 - $32.50
    $32.50
    Pack of 10
       $32.50
    Supplier Number: 0100-1375
  • Reducing nut for split/splitless inlet
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $81.20
    $81.20
    Each
       $81.20
    Supplier Number: 18740-20800
  • Front ferrule, ¹/₄"
    PTFE
    250 °C
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 10
    Pack of 10 - $32.50
    $32.50
    Pack of 10
       $32.50
    Supplier Number: 0100-1378
  • Front ferrule, front ¹/₈"
    Brass
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 10
    Pack of 10 - $31.20
    $31.20
    Pack of 10
       $31.20
    Supplier Number: 5180-4109
  • Ferrule, back, ¹/₈"
    Brass
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 10
    Pack of 10 - $31.20
    $31.20
    Pack of 10
       $31.20
    Supplier Number: 5180-4115
  • Front ferrule, ¹/₁₆", one piece
    Stainless steel
    500 °C
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 10
    Pack of 10 - $92.40
    $92.40
    Pack of 10
       $92.40
    Supplier Number: 5181-1292
  • Column nut adapter, graphite
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $129.00
    $129.00
    Each
       $129.00
    Supplier Number: G3510-20018
  • Ferrule, back, ¹/₁₆", brass
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 10
    Pack of 10 - $36.40
    $36.40
    Pack of 10
       $36.40
    Supplier Number: 5180-4113
  • Ferrule, low thermal mass, 450/530 µm, column Int.Ø 0.45 - 0.53 mm, Int.Ø 0.8 mm, valcon polyimide/graphite, 350 °C
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 5
    Pack of 5 - $150.00
    $150.00
    Pack of 5
       $150.00
    Supplier Number: 5190-1439
  • Column nut for inlet with long or long 2-hole ferrule
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $47.80
    $47.80
    Each
       $47.80
    Supplier Number: 05921-21170
  • Finger-tight column nut, 0.1 to 0.32 mm columns
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $46.50
    $46.50
    Each
       $46.50
    Supplier Number: 5020-8292
  • Finger-tight column nut, 530 µm columns
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $46.50
    $46.50
    Each
       $46.50
    Supplier Number: 5020-8293
  • Blanking plug, finger-tight style
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $37.50
    $37.50
    Each
       $37.50
    Supplier Number: 5020-8294
  • Fly ash filter
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $128.00
    $128.00
    Each
       $128.00
    Supplier Number: 3150-0417
  • Column nut for MS interface
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $32.30
    $32.30
    Each
       $32.30
    Supplier Number: 05988-20066
  • Column nut for GC capillaries
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 2
    Pack of 2 - $70.70
    $70.70
    Pack of 2
       $70.70
    Supplier Number: 5181-8830
  • Insert standard weldment, manual pneumatics
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $542.00
    $542.00
    Each
       $542.00
    Supplier Number: 19251-60575
  • Ferrule/liner kit
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $205.00
    $205.00
    Each
       $205.00
    Supplier Number: 19256-60590
  • Packed port adapter liner, ¹/₈" column
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $219.00
    $219.00
    Each
       $219.00
    Supplier Number: 19243-80510
  • Wrench, open-ended, ¹/₄" and ⁵/₁₆"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $42.30
    $42.30
    Each
       $42.30
    Supplier Number: 8710-0510
  • Ferrule, low thermal mass, column Int.Ø up to 250 µm, 0.4 mm
    Valcon Polyimide/Graphite
    350 °C
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 5
    Pack of 5 - $119.00
    $119.00
    Pack of 5
       $119.00
    Supplier Number: 5190-1437
  • Ferrule, low thermal mass, column Int.Ø 320 µm, 0.5 mm
    Valcon Polyimide/Graphite
    350 °C
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 5
    Pack of 5 - $108.00
    $108.00
    Pack of 5
       $108.00
    Supplier Number: 5190-1438
  • Column nut, for Agilent 6850 GC
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 2
    Pack of 2 - $136.00
    $136.00
    Pack of 2
       $136.00
    Supplier Number: 5183-4732
  • PTV column nut, high temperature, hex
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $355.00
    $355.00
    Each
       $355.00
    Supplier Number: 5188-5312