CrossLab Column Ferrules for Third Party GC Instruments
CrossLab Column Ferrules for Third Party GC Instruments
Certificates
CrossLab Column Ferrules for Third Party GC Instruments
CrossLab Column Ferrules for Third Party GC Instruments
41 Options Available Below
Product Details & Documents
Column ferrules for PerkinElmer, Thermo Scientific, and Varian/Bruker GC Systems.
  • All column ferrules are nonstick and provide a leak-free seal
  • Graphite ferrules provide an easy-to-use and highly stable seal, boast a high temperature limit, and can be removed easily
  • Vespel/graphite ferrules are mechanically robust and maintain effectiveness over a long lifetime
  • Vespel ferrules are mechanically robust, maintain a long lifetime, and can be reused or removed easily
  • To ensure performance and longevity, column ferrules should be replaced at the same time as columns, and when performing column maintenance
Agilent provides a range of high-performance column ferrules for various third-party gas chromatography instruments, including PerkinElmer, Thermo Scientific and Varian/Bruker. Graphite, Vespel/graphite, and Vespel column ferrules create consistent, leak-free seals with minimum torque. All column ferrules accommodate various column outer diameters and can withstand temperature cycling. In addition, the specially designed column ferrules prevent sticking to columns or other fittings. Whatever your application, Agilent's column ferrule portfolio can deliver excellent performance for use with your GC system
Product Family Options
Informations sur le produit
  • Désignation
    Matière
    Ø int.
    Max. temp.
    Availability
    Price
  • Agilent capillary column ferrule, for 0.25 mm columns
    Graphite 15%/Vespel® 85%
    0.4 mm
    350 °C
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 10
    Pack of 10 - $109.00
    $109.00
    Pack of 10
       $109.00
    Supplier Number: 8010-0267
  • Agilent capillary column ferrule, for 0.32 mm columns
    Graphite 15%/Vespel® 85%
    0.5 mm
    350 °C
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 10
    Pack of 10 - $109.00
    $109.00
    Pack of 10
       $109.00
    Supplier Number: 8010-0268
  • Agilent capillary column ferrule, for 0.53 mm columns
    Graphite 15%/Vespel® 85%
    0.8 mm
    350 °C
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 10
    Pack of 10 - $109.00
    $109.00
    Pack of 10
       $109.00
    Supplier Number: 8010-0269
  • Agilent ferrule for PerkinElmer, 2-hole, 0.25 mm Int.Ø column
    Graphite 15%/Vespel® 85%
    0.4 mm
    350 °C
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 10
    Pack of 10 - $171.00
    $171.00
    Pack of 10
       $171.00
    Supplier Number: 8010-0312
  • Agilent ferrule for PerkinElmer, 2-hole, 0.32 mm Int.Ø column
    Graphite 15%/Vespel® 85%
    0.5 mm
    350 °C
    Discontinued
    Supplier Number: 8003-0216
  • Agilent ferrule for Shimadzu, 0.25 mm Int.Ø column
    Graphite
    0.4 mm
    450 °C
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 10
    Pack of 10 - $181.00
    $181.00
    Pack of 10
       $181.00
    Supplier Number: 8001-0211
  • Agilent ferrule for Shimadzu, 0.25 mm Int.Ø column
    Graphite 15%/Vespel® 85%
    0.4 mm
    350 °C
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 10
    Pack of 10 - $139.00
    $139.00
    Pack of 10
       $139.00
    Supplier Number: 8001-0221
  • Agilent ferrule for Shimadzu, 0.32 mm Int.Ø column
    Graphite
    0.5 mm
    450 °C
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 10
    Pack of 10 - $141.00
    $141.00
    Pack of 10
       $141.00
    Supplier Number: 8001-0212
  • Agilent ferrule for Shimadzu, 0.32 mm Int.Ø column
    Graphite 15%/Vespel® 85%
    0.5 mm
    350 °C
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 10
    Pack of 10 - $139.00
    $139.00
    Pack of 10
       $139.00
    Supplier Number: 8001-0222
  • Agilent ferrule for Shimadzu, 0.53 mm Int.Ø column
    Graphite
    0.8 mm
    450 °C
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 10
    Pack of 10 - $141.00
    $141.00
    Pack of 10
       $141.00
    Supplier Number: 8001-0213
  • Agilent ferrule for Shimadzu, 0.53 mm Int.Ø column
    Graphite 15%/Vespel® 85%
    0.8 mm
    350 °C
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 10
    Pack of 10 - $139.00
    $139.00
    Pack of 10
       $139.00
    Supplier Number: 8001-0223
  • Agilent ferrule for Shimadzu, 0.53 mm Int.Ø column
    Graphite 15%/Vespel® 85%
    0.8 mm
    350 °C
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 10
    Pack of 10 - $85.10
    $85.10
    Pack of 10
       $85.10
    Supplier Number: 8010-0313
  • Agilent ferrule for Thermo, 0.25 mm Int.Ø column
    Graphite 15%/Vespel® 85%
    0.4 mm
    350 °C
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 10
    Pack of 10 - $85.10
    $85.10
    Pack of 10
       $85.10
    Supplier Number: 8010-0310
  • Agilent ferrule for Thermo, 0.25 mm Int.Ø or smaller columns
    Graphite 15%/Vespel® 85%
    0.4 mm
    350 °C
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 10
    Pack of 10 - $106.00
    $106.00
    Pack of 10
       $106.00
    Supplier Number: 8002-0220
  • Agilent ferrule for Thermo, 0.32 mm Int.Ø column
    Graphite 15%/Vespel® 85%
    0.5 mm
    350 °C
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 10
    Pack of 10 - $85.10
    $85.10
    Pack of 10
       $85.10
    Supplier Number: 8010-0311
  • Agilent ferrule for Thermo, 0.53 mm Int.Ø column
    Graphite 15%/Vespel® 85%
    0.8 mm
    350 °C
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 10
    Pack of 10 - $106.00
    $106.00
    Pack of 10
       $106.00
    Supplier Number: 8002-0222
  • Agilent ferrule for Thermo, 0.32 mm Int.Ø or smaller columns, ¹/₁₆ fitting size
    Graphite 15%/Vespel® 85%
    0.5 mm
    350 °C
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 10
    Pack of 10 - $112.00
    $112.00
    Pack of 10
       $112.00
    Supplier Number: 8002-0221
  • Agilent ferrule for Varian/Bruker, 0.18 mm Int.Ø or smaller columns
    Vespel®
    0.3 mm
    280 °C
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 10
    Pack of 10 - $83.00
    $83.00
    Pack of 10
       $83.00
    Supplier Number: 8010-0306
  • Agilent ferrule for Varian/Bruker, 0.25 mm Int.Ø column
    Vespel®
    0.4 mm
    280 °C
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 10
    Pack of 10 - $83.00
    $83.00
    Pack of 10
       $83.00
    Supplier Number: 8004-0219
  • Agilent ferrule for Varian/Bruker, 0.25 mm Int.Ø column
    Graphite
    0.4 mm
    450 °C
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 10
    Pack of 10 - $64.20
    $64.20
    Pack of 10
       $64.20
    Supplier Number: 8010-0301
  • Agilent ferrule for Varian/Bruker, 0.25 mm Int.Ø column
    Vespel®
    0.4 mm
    280 °C
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 10
    Pack of 10 - $83.00
    $83.00
    Pack of 10
       $83.00
    Supplier Number: 8010-0307
  • Agilent ferrule for Varian/Bruker, 0.32 mm Int.Ø column
    Graphite
    0.5 mm
    450 °C
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 10
    Pack of 10 - $64.20
    $64.20
    Pack of 10
       $64.20
    Supplier Number: 8010-0302
  • Agilent ferrule for Varian/Bruker, 0.32 mm Int.Ø column
    Vespel®
    0.5 mm
    280 °C
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 10
    Pack of 10 - $83.00
    $83.00
    Pack of 10
       $83.00
    Supplier Number: 8010-0308
  • Agilent ferrule for Varian/Bruker, 0.53 mm Int.Ø column
    Graphite
    0.8 mm
    450 °C
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 10
    Pack of 10 - $61.30
    $61.30
    Pack of 10
       $61.30
    Supplier Number: 8010-0304
  • Agilent ferrule for Varian/Bruker, 0.53 mm Int.Ø column
    Vespel®
    0.8 mm
    280 °C
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 10
    Pack of 10 - $83.00
    $83.00
    Pack of 10
       $83.00
    Supplier Number: 8010-0309
  • Agilent ferrule for Varian/Bruker, 2-hole, 0.25 mm Int.Ø column
    Graphite 40%/Vespel® 60%
    0.4 mm
    400 °C
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 10
    Pack of 10 - $171.00
    $171.00
    Pack of 10
       $171.00
    Supplier Number: 8004-0213
  • Agilent ferrule for Varian/Bruker, 2-hole, 0.32 mm Int.Ø column
    Graphite 40%/Vespel® 60%
    0.5 mm
    400 °C
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 10
    Pack of 10 - $171.00
    $171.00
    Pack of 10
       $171.00
    Supplier Number: 8004-0215
  • Agilent ferrule for Varian/Bruker, 2-hole, 0.32 mm Int.Ø column
    Graphite
    0.5 mm
    450 °C
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 10
    Pack of 10 - $129.00
    $129.00
    Pack of 10
       $129.00
    Supplier Number: 8010-0303
  • Agilent front ferrule for PerkinElmer, ¹/₈"
    Graphite
    3.2 mm
    450 °C
    Discontinued
    Supplier Number: 8003-0212
  • Agilent front ferrule for PerkinElmer, ¹/₈"
    Graphite 15%/Vespel® 85%
    3.2 mm
    350 °C
    Discontinued
    Supplier Number: 8003-0219
  • Agilent front ferrule for Shimadzu, ¹/₁₆"
    Graphite 15%/Vespel® 85%
    1.6 mm
    350 °C
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 10
    Pack of 10 - $85.10
    $85.10
    Pack of 10
       $85.10
    Supplier Number: 8010-0315
  • Agilent front ferrule for Shimadzu, ¹/₄"
    Graphite 15%/Vespel® 85%
    6.35 mm
    350 °C
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 10
    Pack of 10 - $95.70
    $95.70
    Pack of 10
       $95.70
    Supplier Number: 8010-0314
  • Agilent front ferrule for Varian/Bruker, ¹/₄", straight bExt.Øy
    Graphite
    6.35 mm
    450 °C
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 10
    Pack of 10 - $68.20
    $68.20
    Pack of 10
       $68.20
    Supplier Number: 8010-0305
  • Agilent straight ferrule for PerkinElmer, ¹/₄"
    Vespel®
    6.35 mm
    280 °C
    Discontinued
    Supplier Number: 8003-0223
  • Agilent straight ferrule for Thermo, 0.25 mm Int.Ø or smaller columns
    Graphite
    0.4 mm
    450 °C
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 10
    Pack of 10 - $60.20
    $60.20
    Pack of 10
       $60.20
    Supplier Number: 8002-0215
  • Agilent straight ferrule for Thermo, 0.32 mm Int.Ø or smaller columns
    Graphite
    0.5 mm
    450 °C
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 10
    Pack of 10 - $60.20
    $60.20
    Pack of 10
       $60.20
    Supplier Number: 8002-0216
  • Agilent straight ferrule for Thermo, 0.53 mm Int.Ø or smaller columns
    Graphite
    0.8 mm
    350 °C
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 10
    Pack of 10 - $60.20
    $60.20
    Pack of 10
       $60.20
    Supplier Number: 8002-0217
  • Agilent straight ferrule for Thermo, for M4 fittings, 0.25 mm Int.Ø or smaller columns
    Graphite
    0.4 mm
    450 °C
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 10
    Pack of 10 - $185.00
    $185.00
    Pack of 10
       $185.00
    Supplier Number: 8002-0212
  • Agilent straight ferrule for Thermo, for M4 fittings, 0.32 mm Int.Ø or smaller columns
    Graphite
    0.5 mm
    450 °C
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 10
    Pack of 10 - $185.00
    $185.00
    Pack of 10
       $185.00
    Supplier Number: 8002-0213
  • Agilent straight ferrule for Thermo, for M4 fittings, 0.53 mm Int.Ø or smaller columns
    Graphite
    0.8 mm
    450 °C
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 10
    Pack of 10 - $185.00
    $185.00
    Pack of 10
       $185.00
    Supplier Number: 8002-0214
  • Agilent straight ferrule for Varian/Bruker, 0.18 mm Int.Ø, or smaller column
    Graphite 40%/Vespel® 60%
    0.3 mm
    400 °C
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 10
    Pack of 10 - $85.10
    $85.10
    Pack of 10
       $85.10
    Supplier Number: 8004-0211
  • Agilent straight ferrule for Varian/Bruker, 0.25 mm id column
    Graphite 40%/Vespel® 60%
    0.4 mm
    400 °C
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 10
    Pack of 10 - $85.10
    $85.10
    Pack of 10
       $85.10
    Supplier Number: 8004-0212
  • Agilent straight ferrule for Varian/Bruker, 0.32 mm Int.Ø column
    Graphite 40%/Vespel® 60%
    0.5 mm
    400 °C
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 10
    Pack of 10 - $85.10
    $85.10
    Pack of 10
       $85.10
    Supplier Number: 8004-0214
  • Agilent straight ferrule for Varian/Bruker, 0.53 mm Int.Ø column
    Graphite 40%/Vespel® 60%
    0.8 mm
    400 °C
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 10
    Pack of 10 - $85.10
    $85.10
    Pack of 10
       $85.10
    Supplier Number: 8004-0216
  • Agilent straight ferrule for Varian/Bruker, ¹/₄", straight bExt.Øy
    Graphite 40%/Vespel® 60%
    6.4 mm
    400 °C
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 10
    Pack of 10 - $95.70
    $95.70
    Pack of 10
       $95.70
    Supplier Number: 8004-0217