Detectors for ICP-MS, Agilent Technologies
High quality, long lifetime electron multiplier detector for ICP-MS 7700, 7800, 7900, 8800, and 8900.
- Long lifetime pulse - analogue detector provides superior wide linear dynamic range of up to 11 orders to allow measurement of major and trace elements in a single run
- Electron multiplier converts incident ions into an amplified electric current in a stable, precision process
- The low noise currents created are used to measure the ion signal from sample in a wide variety of ICP-MS applications.
- As voltage increases, the amplification factor (also known as gain) increases in the system
Long lifetime pulse - analogue detector provides superior wide linear dynamic range of up to 11 orders to allow measurement of major and trace elements in a single run;The Agilent high quality electron multiplier for ICP-MS converts incident ions into an amplified electric current used to record measurements of the ion signal. As the voltage in the system increases the amplification factor (or gain) increases.
Frequently Bought Together