CrossLab Rotating Cylinder for Dissolution, Agilent Technologies
Catalog # CAAG12-1370
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
Specifications
- Description:Cuprophan membrane, 126×345 mm, 10 sheets/pack
- Cat. no.:CAAG12-1370
- Certification:No
Specifications
About this item
Uses a stainless steel cylinder and includes two sizes for smaller and larger patches.
- No need to match an upper and lower assembly
The rotating cylinder for USP Apparatus 6 assembly uses a stainless steel cylinder and includes four slots at the top for improved media circulation within the dissolution vessel. Each rotating cylinder includes two sizes, one for smaller patches, the other for larger ones.