CrossLab Reciprocating Dissolution Holders, Agilent Technologies | Avantor
CrossLab Reciprocating Dissolution Holders, Agilent Technologies
CrossLab Reciprocating Dissolution Holders, Agilent Technologies
Certificates
CrossLab Reciprocating Dissolution Holders, Agilent Technologies
CrossLab Reciprocating Dissolution Holders, Agilent Technologies
31 Options Available Below
Product Details & Documents
Agilent can provide many different dosage form holders for the Apparatus 7.
  • Accessories for the reciprocating holder apparatus 7
  • Simulates the biorelevant conditions for the skin, gastrointestinal tract and tissue required for transdermal, oral drugs and implants with biorelevant temperature agitation rates and retention times
  • Accommodates a variety of dosage form holders including reciprocating disks, angled disks, spring holders, stent holders, and pointed rods
  • Supports typical volume configurations of 5 to 300 ml, depending on instrument
Ideal for automatic dissolution testing of dosage forms that require a change of media, a smaller volume, or more vigorous agitation. The Agilent 400-DS dissolution apparatus 7 enables of small-volume drug-release testing of medical devices such as drug eluting stents (DES).
Product Family Options
Informations sur le produit
  • Désignation
    Availability
    Price
  • 50-mesh Basket holder Assembly, stainless steel, use with 10 ml cell, for 400-DS
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $2,850.00
    $2,850.00
    Each
       $2,850.00
    Supplier Number: 33-9041
  • 50-mesh Basket holder Assembly, stainless steel, use with 5 ml cell, for 400-DS
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,740.00
    $1,740.00
    Each
       $1,740.00
    Supplier Number: 33-9052
  • 50-mesh Basket, stainless steel, use with 10 ml cell, for 400-DS
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $810.00
    $810.00
    Each
       $810.00
    Supplier Number: 33-9029
  • 50-mesh Basket, stainless steel, use with 5 ml cell, for 400-DS
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $821.00
    $821.00
    Each
       $821.00
    Supplier Number: 33-9051
  • Accessory Kit, 1.7 mm Holder for Pacemaker Lead, for 400-DS
    Log in pour la disponibilité
    Log in pour la disponibilité
    Kit of 1
    Kit of 1 - $453.00
    $453.00
    Kit of 1
       $453.00
    Supplier Number: 33-9025
  • Angled Disk Holder 1.42", for BIO-DIS
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $723.00
    $723.00
    Each
       $723.00
    Supplier Number: 27-8036
  • Basket Assembly, Titanium, for BIO-DIS
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $2,270.00
    $2,270.00
    Each
       $2,270.00
    Supplier Number: 27-8622
  • Basket,Mesh End, 20 µm, for 400-DS
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,930.00
    $1,930.00
    Each
       $1,930.00
    Supplier Number: 33-9046
  • Contact Lens Holder Assembly, use with 5 ml cell, for 400-DS
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,580.00
    $1,580.00
    Each
       $1,580.00
    Supplier Number: 33-9048
  • Extraction Tool, for 400-DS
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $69.80
    $69.80
    Each
       $69.80
    Supplier Number: 33-9007
  • Mini basket shaft, threaded, for BIO-DIS
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,070.00
    $1,070.00
    Each
       $1,070.00
    Supplier Number: 27-8600
  • Mini basket, 40-mesh, for BIO-DIS
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,900.00
    $1,900.00
    Each
       $1,900.00
    Supplier Number: 27-8620
  • Mini basket, 50-mesh, for BIO-DIS
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,900.00
    $1,900.00
    Each
       $1,900.00
    Supplier Number: 27-8621
  • PEEK Basket holder Assembly, for 400-DS
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,620.00
    $1,620.00
    Each
       $1,620.00
    Supplier Number: 33-9022
  • Pointed acrylic rod kit, for BIO-DIS
    Log in pour la disponibilité
    Log in pour la disponibilité
    Kit of 1
    Kit of 1 - $839.00
    $839.00
    Kit of 1
       $839.00
    Supplier Number: 27-3000
  • Pointed acrylic rod replacement, for BIO-DIS
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $453.00
    $453.00
    Each
       $453.00
    Supplier Number: 27-3002
  • Reciprocating Disk, Alza Type, 1.6 cm sq., for BIO-DIS
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $669.00
    $669.00
    Each
       $669.00
    Supplier Number: 27-8005
  • Reciprocating Disk, Alza Type, 7.0 cm sq., for BIO-DIS
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $839.00
    $839.00
    Each
       $839.00
    Supplier Number: 27-8020
  • Spring holder, Alza, 0.60" L × 0.25" int.Ø × 0.031" wire int.Ø, spring centered, for BIO-DIS
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $502.00
    $502.00
    Each
       $502.00
    Supplier Number: 27-0108
  • Spring holder, Alza, 0.60" L × 0.25" int.Ø × 0.031" wire int.Ø, spring offset, for BIO-DIS
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $501.00
    $501.00
    Each
       $501.00
    Supplier Number: 27-0103
  • Spring holder, Alza, 0.96" L × 0.33" int.Ø × 0.031" wire int.Ø, spring centered, for BIO-DIS
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $564.00
    $564.00
    Each
       $564.00
    Supplier Number: 27-0107
  • Spring holder, Alza, 0.96" L × 0.33" int.Ø × 0.031" wire int.Ø, spring offset, for BIO-DIS
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $445.00
    $445.00
    Each
       $445.00
    Supplier Number: 27-0102
  • Spring holder, Alza, 1.00" L × 0.50" int.Ø × 0.031" wire int.Ø, spring centered, for BIO-DIS
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,180.00
    $1,180.00
    Each
       $1,180.00
    Supplier Number: 27-0109
  • Spring holder, Alza, 1.00" L × 0.50" int.Ø × 0.031" wire int.Ø, spring offset, for BIO-DIS
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,030.00
    $1,030.00
    Each
       $1,030.00
    Supplier Number: 27-0104
  • Spring holder, Alza, 1.40" L × 0.31" int.Ø × 0.040" wire int.Ø, spring centere, for BIO-DIS
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $502.00
    $502.00
    Each
       $502.00
    Supplier Number: 27-0106
  • Spring holder, Alza, 1.40" L × 0.31" int.Ø × 0.040" wire int.Ø, spring offset, for BIO-DIS
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $445.00
    $445.00
    Each
       $445.00
    Supplier Number: 27-0101
  • Spring holder, Alza, 1.45" L × 0.58" int.Ø × 0.031" wire int.Ø, spring centered, for BIO-DIS
    Discontinued
    Supplier Number: -
  • Spring holder, Alza, 1.45" L × 0.58" int.Ø × 0.031" wire int.Ø, spring offset, for BIO-DIS
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $501.00
    $501.00
    Each
       $501.00
    Supplier Number: 27-0100
  • Stent Holder Assembly, use with 10 ml cell, for 400-DS
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,210.00
    $1,210.00
    Each
       $1,210.00
    Supplier Number: 33-9012
  • Stent Holder Assembly, use with 5 ml cell, for 400-DS
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $941.00
    $941.00
    Each
       $941.00
    Supplier Number: 33-9009
  • Temperature Calibration Assembly, use with 10 ml cell, for 400-DS
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $928.00
    $928.00
    Each
       $928.00
    Supplier Number: 33-9018
  • Transdermal patch holder kit, for BIO-DIS
    Log in pour la disponibilité
    Log in pour la disponibilité
    Kit of 1
    Kit of 1 - $1,340.00
    $1,340.00
    Kit of 1
       $1,340.00
    Supplier Number: 27-3001