CrossLab Dissolution Shafts, Agilent Technologies
CrossLab Dissolution Shafts, Agilent Technologies
Certificates
CrossLab Dissolution Shafts, Agilent Technologies
CrossLab Dissolution Shafts, Agilent Technologies
12 Options Available Below
Product Details & Documents
Shafts of different materials and lengths, individually serialized to meet standard USP design.
  • Electropolished stainless steel, PTFE-coated, gold-plated or mini basket shafts available
  • Individually serialized
  • Three-clip shafts to meet standard USP design
  • O-ring shafts for automated systems
  • Conical shafts to support air bubble dispersion
  • A range of basket shaft lengths are available to accommodate various models and volume configurations
Agilent provides stainless steel, PTFE-coated, gold-plated, or mini shafts that are individually serialized. Several basket and paddle shaft lengths and options are available to meet standard USP design.
Product Family Options
Informations sur le produit
  • Désignation
    Certification
    Availability
    Price
  • Basket shaft, 3‐clip, PTFE-coated, 21", for 705-DS; 708-DS; 709-DS; VK7000
    Yes
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $891.00
    $891.00
    Each
       $891.00
    Supplier Number: 13-3622
    Spécifications:
    More
    Specifications
    Cat. No.
    CAAG13-3622
  • Basket shaft, electropolished stainless steel Spin on/off, O-ring, 21", for 705-DS; 708-DS; 709-DS; VK7000
    Yes
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $484.00
    $484.00
    Each
       $484.00
    Supplier Number: 13-3632
    Spécifications:
    More
    Specifications
    Cat. No.
    CAAG13-3632
  • Basket Shaft, electropolished stainless steel, 3-clip, Conical, 21", for 705-DS; 708-DS; 709-DS; VK7000
    Yes
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $663.00
    $663.00
    Each
       $663.00
    Supplier Number: 14-3624
    Spécifications:
    More
    Specifications
    Cat. No.
    CAAG14-3624
  • Basket shaft, electropolished stainless steel, O-ring, for Bolus Basket, 24", for 705-DS; 708-DS; 709-DS; VK7000
    Yes
    Discontinued
    Supplier Number: 13-3630
    Spécifications:
    More
    Specifications
    Cat. No.
    CAAG13-3630
  • Basket shaft, electropolished stainless steel, USP 3-clip, 21", for 705-DS; 708-DS; 709-DS; VK7000
    Yes
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $452.00
    $452.00
    Each
       $452.00
    Supplier Number: 14-3620
    Spécifications:
    More
    Specifications
    Cat. No.
    CAAG14-3620
  • Basket shaft, electropolished stainless steel, USP 3‐clip, 24", for 705-DS; 708-DS; 709-DS; VK7000
    Yes
    Discontinued
    Supplier Number: 13-3629
    Spécifications:
    More
    Specifications
    Cat. No.
    CAAG13-3629
  • Basket shaft, lower interchangeable Spin on/off, electropolished stainless steel, 3-clip, for 2 L vessels, 21", for 705-DS; 708-DS; 709-DS; VK7000
    Yes
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $616.00
    $616.00
    Each
       $616.00
    Supplier Number: 14-3634
    Spécifications:
    More
    Specifications
    Cat. No.
    CAAG14-3634
  • Basket shaft, lower interchangeable Spin on/off, electropolished stainless steel, 3-clip, USP, 21", for 705-DS; 708-DS; 709-DS; VK7000
    Yes
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $379.00
    $379.00
    Each
       $379.00
    Supplier Number: 14-3631
    Spécifications:
    More
    Specifications
    Cat. No.
    CAAG14-3631
  • Basket shaft, lower interchangeable Spin on/off, electropolished stainless steel, 3-clip, USP, Pack of 6, 21", for 705-DS; 708-DS; 709-DS; VK7000
    Yes
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $2,180.00
    $2,180.00
    Each
       $2,180.00
    Supplier Number: 16-3631
    Spécifications:
    More
    Specifications
    Cat. No.
    CAAG16-3631
  • Basket shaft, USP 3‐clip, 14.5", for VK7025/7030
    Yes
    Discontinued
    Supplier Number: 13-3615
    Spécifications:
    More
    Specifications
    Cat. No.
    CAAG13-3615
  • Mini basket shaft, lower interchangeable Spin on/off, electropolished stainless steel, O-ring, 21", for 705-DS; 708-DS; 709-DS; VK7000
    Yes
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $499.00
    $499.00
    Each
       $499.00
    Supplier Number: 14-3636
    Spécifications:
    More
    Specifications
    Cat. No.
    AG14-3636
  • Upper Receptor Shaft, Spin on/off, electropolished stainless steel, 14.75", for 705-DS; 708-DS; 709-DS; VK7000
    Yes
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $379.00
    $379.00
    Each
       $379.00
    Supplier Number: 14-3613
    Spécifications:
    More
    Specifications
    Cat. No.
    CAAG14-3613
  • Upper Receptor Shaft, Spin on/off, electropolished stainless steel, 14.75", Pack of 6, for 705-DS; 708-DS; 709-DS; VK7000
    Yes
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $2,180.00
    $2,180.00
    Each
       $2,180.00
    Supplier Number: 16-3613
    Spécifications:
    More
    Specifications
    Cat. No.
    CAAG16-3613
  • Upper Receptor Shaft, Spin on/off, electropolished stainless steel, 17.75", for 705-DS; 708-DS; 709-DS; VK7000
    Yes
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $575.00
    $575.00
    Each
       $575.00
    Supplier Number: 13-3614
    Spécifications:
    More
    Specifications
    Cat. No.
    CAAG13-3614
  • Upper Receptor Shaft, Spin on/off, electropolished stainless steel, 8.75", for VK7030
    Yes
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $483.00
    $483.00
    Each
       $483.00
    Supplier Number: 14-3612
    Spécifications:
    More
    Specifications
    Cat. No.
    CAAG14-3612