CrossLab Dissolution Paddles, Agilent Technologies | Avantor
CrossLab Dissolution Paddles, Agilent Technologies
CrossLab Dissolution Paddles, Agilent Technologies
Certificates
CrossLab Dissolution Paddles, Agilent Technologies
CrossLab Dissolution Paddles, Agilent Technologies
16 Options Available Below
Product Details & Documents
Available in stainless steel, PTFE-coated, or PEEK in a range of sizes.
  • All paddles are serialized and designed to meet USP specifications for Apparatus 2
  • Select from electropolished stainless steel, PTFE-coated, or the biocompatible, chemically inert PEEK paddles in a range of sizes
  • Additionally, mini and mega paddles for small- and large-volume dissolution testing are available
All paddles are serialized and designed to meet USP specifications for USP Apparatus 2 including an individual CoC as standard. Paddles are available in stainless steel, PTFE-coated, or PEEK in a range of standard, mega, or mini sizes.
Certifications: Each paddle includes an individual Certificate of Conformance as standard.
Product Family Options
Informations sur le produit
  • Désignation
    Certification
    Availability
    Price
  • Mini Paddle, electropolished stainless steel, 14.5", for VK7030
    Yes
    Discontinued
    Supplier Number: 13-3593
  • Mini paddle, electropolished stainless steel, 24", for 705-DS; 708-DS; 709-DS; VK7000
    Yes
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $977.00
    $977.00
    Each
       $977.00
    Supplier Number: 14-3600
  • Mini paddle, lower interchangeable Spin on/off, electropolished stainless steel, 21", for 705-DS; 708-DS; 709-DS; VK7000
    Yes
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $632.00
    $632.00
    Each
       $632.00
    Supplier Number: 14-3608
  • Mini paddle, lower interchangeable Spin on/off, PTFE-coated, 21", for 705-DS; 708-DS; 709-DS; VK7000
    Yes
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $796.00
    $796.00
    Each
       $796.00
    Supplier Number: 14-3607
  • Mini Paddle, PTFE-coated, 14.5", for VK7030
    Yes
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $693.00
    $693.00
    Each
       $693.00
    Supplier Number: 13-3592
  • Mini paddle, PTFE-coated, 24", for 705-DS; 708-DS; 709-DS; VK7000
    Yes
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,110.00
    $1,110.00
    Each
       $1,110.00
    Supplier Number: 14-3599
  • Paddle, electropolished stainless steel, 14.5", for VK7030
    Yes
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $616.00
    $616.00
    Each
       $616.00
    Supplier Number: 14-3590
  • Paddle, electropolished stainless steel, 21", for 705-DS; 708-DS; 709-DS; VK7000
    Yes
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $533.00
    $533.00
    Each
       $533.00
    Supplier Number: 14-3595
  • Paddle, electropolished stainless steel, 24", for 705-DS; 708-DS; 709-DS; VK7000
    Yes
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $813.00
    $813.00
    Each
       $813.00
    Supplier Number: 14-3597
  • Paddle, lower interchangeable Spin on/off, electropolished stainless steel, 1 L, 21", for 705-DS; 708-DS; 709-DS; VK7000
    Yes
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $419.00
    $419.00
    Each
       $419.00
    Supplier Number: 14-3603
  • Paddle, lower interchangeable Spin on/off, electropolished stainless steel, 1 L, pack of 6, 21", for 705-DS; 708-DS; 709-DS; VK7000
    Yes
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $2,390.00
    $2,390.00
    Each
       $2,390.00
    Supplier Number: 16-3603
  • Paddle, lower interchangeable Spin on/off, electropolished stainless steel, 2 L, 21", for 705-DS; 708-DS; 709-DS; VK7000
    Yes
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $566.00
    $566.00
    Each
       $566.00
    Supplier Number: 14-3605
  • Paddle, lower interchangeable Spin on/off, PEEK, for 1 L, 21", for 705-DS; 708-DS; 709-DS; VK7000
    Yes
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $960.00
    $960.00
    Each
       $960.00
    Supplier Number: 14-3606
  • Paddle, lower interchangeable Spin on/off, PTFE-coated, 1 L, 21", for 705-DS; 708-DS; 709-DS; VK7000
    Yes
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $458.00
    $458.00
    Each
       $458.00
    Supplier Number: 14-3602
  • Paddle, lower interchangeable Spin on/off, PTFE-coated, 1 L, pack of 6, 21", for 705-DS; 708-DS; 709-DS; VK7000
    Yes
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $2,720.00
    $2,720.00
    Each
       $2,720.00
    Supplier Number: 16-3602
  • Paddle, lower interchangeable Spin on/off, PTFE-coated, for 2 L, 21", for 705-DS; 708-DS; 709-DS; VK7000
    Yes
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $960.00
    $960.00
    Each
       $960.00
    Supplier Number: 14-3604
  • Paddle, PTFE-coated, 21", for 705-DS; 708-DS; 709-DS; VK7000
    Yes
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $730.00
    $730.00
    Each
       $730.00
    Supplier Number: 14-3594
  • Paddle, PTFE-coated, 24", for 705-DS; 708-DS; 709-DS; VK7000
    Yes
    Discontinued
    Supplier Number: 13-3596