CrossLab Dissolution Full Flow Filters, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
Certificates
About this item
Made of either color-coded UHMWPE or PVDF, the full flow filter prevents clogging.
- Available in ultrahigh molecular weight polyethylene (UHMWPE) or polyvinylidene fluoride (PVDF)
- Full flow filter offers increased surface area to prevent clogging
- Low protein-binding PVDF filter material available
- Usable with ¹/₈" diameter cannulas
- Available in packs of 100 or 1000
- Full flow filters are color coded for easy identification
- Certificates of analysis can be provided on request
- Standard conventional filter tips are made of polyethylene fluoride and can also be used with ¹/₈" diameter cannulas
Made of either color-coded UHMWPE or PVDF, the full flow filter offers increased surface area to optimize filter life and prevent clogging.
Frequently Bought Together