InfinityLab Well Plates, Agilent Technologies
InfinityLab Well Plates, Agilent Technologies
InfinityLab Well Plates, Agilent Technologies
InfinityLab Well Plates, Agilent Technologies
InfinityLab Well Plates, Agilent Technologies
A comprehensive selection of InfinityLab well plates and sealing mats for the HPLC workflow.
  • Ensure a perfect fit between well plates and slitted sealing mats
  • Access the right formats to ensure each well plate sampler setup is perfectly equipped for all HPLC applications
  • Optimized for high performance in your chromatography workflow
  • A large range of well volumes and package quantities provide the flexibility needed to suit your lab size and your needs
  • Compliant with worldwide ANSI and SLAS standards
Agilent provides a comprehensive selection of InfinityLab well plates and sealing mats for the HPLC workflow. 96 and 384 plates are available in different well sizes, well shapes, and bottom shapes to best suit your needs for all possible applications.
Choosing the right well plate can help you avoid such pitfalls as missed data points, sample rerun, and a failure to meet project deadlines. Agilent InfinityLab well plates and sealing mats for the HPLC workflow will help to produce the highest quality data for most common screening challenges.
Product Family Options
  • Désignation
    Matière
    Well Volume
    Hauteur
    Bottom Style
    Well Shape
  • 96-well plate
    PP
    0.19 ml
    22 mm
    Square
    Pack of 25
    Pack of 25 - $310.00
    $310.00
    Pack of 25
       $310.00
     : 5043-9315
  • 96-well plate
    PP
    0.33 ml
    14 mm
    U
    Round
    Pack of 25
    Pack of 25 - $97.80
    $97.80
    Pack of 25
       $97.80
     : 5043-9312
  • 96-well plate
    PP
    0.33 ml
    14 mm
    V
    Round
    Pack of 50
    Pack of 50 - $186.00
    $186.00
    Pack of 50
       $186.00
     : 5043-9313
  • 96-well plate
    PP
    0.33 ml
    14 mm
    V
    Round
    Pack of 100
    Pack of 100 - $352.00
    $352.00
    Pack of 100
       $352.00
     : 5043-9314
  • 96-well plates with glass inserts, caps, septa preassembled
    PP
    0.35 ml
    U
    Each
    Each - $158.00
    $158.00
    Each
       $158.00
     : 5065-4402
  • 96-well plate
    PP
    0.5 ml
    14 mm
    U
    Round
    Pack of 30
    Pack of 30 - $133.00
    $133.00
    Pack of 30
       $133.00
     : 5043-9310
  • 96-well plate
    PP
    0.5 ml
    14 mm
    U
    Round
    Pack of 120
    Pack of 120 - $446.00
    $446.00
    Pack of 120
       $446.00
     : 5043-9311
  • 96-well plate
    PP
    1.0 ml
    32 mm
    U
    Round
    Pack of 50
    Pack of 50 - $831.00
    $831.00
    Pack of 50
       $831.00
     : 5043-9305
  • 96-well plate
    PP
    1.2 ml
    27 mm
    V
    Round
    Pack of 25
    Pack of 25 - $212.00
    $212.00
    Pack of 25
       $212.00
     : 5043-9308
  • 96-well plate
    PP
    1.2 ml
    27 mm
    V
    Round
    Pack of 50
    Pack of 50 - $404.00
    $404.00
    Pack of 50
       $404.00
     : 5043-9309
  • 96-well plate
    PP
    2.0 ml
    45 mm
    U
    Round
    Pack of 30
    Pack of 30 - $372.00
    $372.00
    Pack of 30
       $372.00
     : 5043-9302
  • 96-well plate
    PP
    2.2 ml
    41 mm
    U
    Square
    Pack of 30
    Pack of 30 - $282.00
    $282.00
    Pack of 30
       $282.00
     : 5043-9300
Sealing Mats
  •  
    Désignation
    Matière
  • Closing mats for 96 well plates
    Silicone
    Pack of 50
    Pack of 50 - $570.00
    $570.00
    Pack of 50
       $570.00
  •  
    Désignation
    Matière
  • Sealing mat, 96 wells, square, preslitted
    Silicone
    Pack of 50
    Pack of 50 - $547.00
    $547.00
    Pack of 50
       $547.00
  •  
    Désignation
    Matière
  • Sealing mat, 96 wells, round, preslitted
    Silicone
    Pack of 100
    Pack of 100 - $1,050.00
    $1,050.00
    Pack of 100
       $1,050.00
  •  
    Désignation
    Matière
  • Sealing mat, 384 wells, square, preslitted
    Silicone
    Pack of 50
    Pack of 50 - $619.00
    $619.00
    Pack of 50
       $619.00
  •  
    Désignation
    Matière
  • Sealing mat, 96 wells, round, preslitted
    Silicone
    Pack of 50
    Pack of 50 - $547.00
    $547.00
    Pack of 50
       $547.00
Accessories
  •  
    Description
  • CAAG5188-5321
    Glass inserts, 350 µl
    Pack of 1,000 - $424.00
  •  
    Description
  • CAAG5188-5322
    Cap for 350 µl glass insert
    Pack of 1,000 - $1,200.00