CrossLab Solvent Inlet Tubing and Filters for HPLC, Agilent Technologies
New Product
High-quality glass and stainless steel solvent filters with different pore sizes for LC applications.
- Solvent inlet filters represent the first barrier for retaining particles, precipitates, and microbes from mobile phases, buffers, and salt solutions
- Filters are significant elements for preventing system blockage, pressure increases, and contamination
- Agilent solvent filters have a defined, homogeneous pore size to effectively block particles above a certain size, while letting the mobile phase through without a significant pressure increase
Agilent provides a comprehensive range of high-quality glass and stainless steel solvent filters with different pore sizes, suitable for many different LC applications and equipment, to optimize HPLC results by removing any trace of particles from the solvent.
All solvent filters and frit adapters are manufactured to Agilent's high levels of quality and have been tested on Agilent instruments to ensure performance that will significantly increase your analytical, instrument, and laboratory efficiency.
Frequently Bought Together