CrossLab Sample Loops for HPLC, Agilent Technologies
A range of high-quality loop capillaries suitable for use with a variety of autosampling equipment.
- Agilent LC metering device Supplies are designed to operate with your LC instruments to work together seamlessly for maximum efficiency and performance–regardless of application area
- Supporting your efforts to increase your analytical, instrument, and laboratory efficiency in your everyday work
Agilent manufactures a range of high-quality loop capillaries suitable for use with a variety of autosampling equipment. Loop capacities vary from 8 µl up to 900 µl; an even larger capacity 5 ml model also available.
Agilent offers both stainless steel and PEEK loops for different pressure ranges and applications.
All loops are are manufactured to exacting quality standards and designed to deliver robust, high-precision performance in various Agilent autosamplers.
Frequently Bought Together