CrossLab Alignment Interfaces, Agilent Technologies
Fournisseur: AGILENT TECHNOLOGIES, INC (CSD) CA
Certificats
À propos de cet article
Together with the capillary cassette, alignment interfaces simplify capillary exchange.
- The quick-change cassette allows capillary exchange in less than a minute
- The cassette and interfaces accept all commercially available capillaries with a 365 µm Ext. Ø
Agilent alignment interfaces, with the capillary cassette, simplify capillary exchange, protect the fragile detection window, and ensure exact alignment of the window in the detector. Alignment interfaces are an integral part of the Agilent diode-array detection (DAD) system. These interfaces contain optical slits that are precisely matched to the capillary inner diameter for optimized sensitivity and linear detection range.
Frequently Bought Together