VWR® Shell Vial and Plug Cap
Shell vials with plug closure, 1.00 ml, Flat bottom, 8×40 mm, Borosilicate glass, Clear, Oui, PE
Supplier: VWR International
Jump to:
Product Details & Documents
Designed for use in Alcott®, Waters®, Gilson®, or Shimadzu® autosamplers, the clear snap-plug closures of these shell vials can be pierced with standard autosampler syringes without vacuum formation.
- Polyethylene plug with starburst center design eases syringe needle penetration
- Convenient vial kits include equal quantities of vials and caps
- Provides an economical option to autosampler vials
Manufactured from borosilicate glass. Vials and snap-plug closures packed separately in clear-pack, three chamber trays.
Spécifications
- Avec bouchon:Oui
- Cap Material:PE
- Capacité:1.00 ml
- Color:Clear
- Désignation:Shell vials with plug closure
- Matière:Borosilicate glass
- Diameter ×H:8×40 mm
- Bottom Style:Flat bottom